Please login to your online account to display your discounted pricing

DL-4-Fluorophenylalanine 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  119580010

 View more versions of this product

Catalog No. AC119580010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 183.18
Quantity 1g
Color White to off-white
EINECS Number 200-113-7
TSCA TSCA
RTECS Number AY6315000
Beilstein 14, IV, 1673
Chemical Name or Material DL-4-Fluorophenylalanine
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H10FNO2
Linear Formula FC6H4CH2CH(NH2)CO2H
MDL Number MFCD00002600
CAS 51-65-0
Packaging Glass bottle

Specifications

Chemical Name or Material DL-4-Fluorophenylalanine
Name Note 97%
CAS 51-65-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10FNO2
Linear Formula FC6H4CH2CH(NH2)CO2H
Formula Weight 183.18
MDL Number MFCD00002600