Please login to your online account to display your discounted pricing

DL-4-Fluorophenylalanine 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material DL-4-Fluorophenylalanine
Name Note 97%
CAS 51-65-0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C9H10FNO2
Linear Formula FC6H4CH2CH(NH2)CO2H
Formula Weight 183.18g/mol
MDL Number MFCD00002600
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119580010

  SDS
fisher scientific acros
119580010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color White to off-white
Chemical Name or Material DL-4-Fluorophenylalanine
Name Note 97%
CAS 51-65-0
Physical Form White to off-white powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C9H10FNO2
Linear Formula FC6H4CH2CH(NH2)CO2H
Formula Weight 183.18g/mol
MDL Number MFCD00002600
Beilstein 14, IV, 1673
RTECS Number AY6315000
EINECS Number 200-113-7
TSCA TSCA