Please login to your online account to display your discounted pricing

DL-4-Fluoro-alpha-methylbenzylamine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material DL-4-Fluoro-α-methylbenzylamine
Name Note 97%
CAS 403-40-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H10FN
Linear Formula FC6H4CH(CH3)NH2
Formula Weight 139.17
MDL Number MFCD00041323
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC167380010

  SDS
thermo fisher scientific
AC167380010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC167380050

  SDS
thermo fisher scientific
167380050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 139.17
Physical Form Liquid
Quantity 1g
Color Clear colorless to yellow or light orange
EINECS Number 206-958-8
Density 1.0590g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5005 to 1.5025 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 403-40-7
Name Note 97%
Linear Formula FC6H4CH(CH3)NH2
MDL Number MFCD00041323
Assay Percent Range 97%
Molecular Formula C8H10FN
Chemical Name or Material DL-4-Fluoro-α-methylbenzylamine
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.