missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-4-Fluoro-alpha-methylbenzylamine, 97%, ACROS Organics™

Manufacturer:  Acros Organics 167380050

 View more versions of this product

Catalog No. AC167380050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

DL-4-Fluoro-α-methylbenzylamine
Authentic
1.5005 to 1.5025 (20°C, 589nm)
5g
403-40-7
C8H10FN
MFCD00041323
QGCLEUGNYRXBMZ-UHFFFAOYSA-N
1-(4-fluorophenyl)ethanamine
123063
Liquid
1.0590g/mL
Glass bottle
Colorless to Yellow or Orange
97%
97%
FC6H4CH(CH3)NH2
1-4-fluorophenyl ethanamine, 1-4-fluorophenyl ethylamine, 4-fluoro-alpha-methylbenzylamine, 1-4'-fluorophenyl ethanamine, dl-4-fluoro-alpha-methylbenzylamine, 4-fluoro-a-methylbenzylamine, 1-p-fluorophenyl-ethylamine, 1-4-fluorophenyl ethan-1-amine, p-fluoro-a-methylbenzylamine, rs-1-4-fluorophenyl ethylamine
CC(C1=CC=C(C=C1)F)N
139.173
139.17
≥96.0%

Chemical Identifiers

403-40-7
139.173
QGCLEUGNYRXBMZ-UHFFFAOYSA-N
123063
CC(C1=CC=C(C=C1)F)N
C8H10FN
MFCD00041323
1-4-fluorophenyl ethanamine, 1-4-fluorophenyl ethylamine, 4-fluoro-alpha-methylbenzylamine, 1-4'-fluorophenyl ethanamine, dl-4-fluoro-alpha-methylbenzylamine, 4-fluoro-a-methylbenzylamine, 1-p-fluorophenyl-ethylamine, 1-4-fluorophenyl ethan-1-amine, p-fluoro-a-methylbenzylamine, rs-1-4-fluorophenyl ethylamine
1-(4-fluorophenyl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-958-8

SDS Documents