missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-4-Chlorophenylalanine 98%, ACROS Organics™

$148.93 - $148.93

Chemical Identifiers

CAS 7424-00-2
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.634
MDL Number MFCD00002601
InChI Key NIGWMJHCCYYCSF-UHFFFAOYSA-N
Synonym fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro
PubChem CID 4652
IUPAC Name 2-amino-3-(4-chlorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC157280050 SDS View Documents Acros Organics
157280050
5g Glass bottle Each for $148.93
AC157280250 SDS View Documents Acros Organics
157280250
25g Plastic bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7424-00-2
199.634
NIGWMJHCCYYCSF-UHFFFAOYSA-N
4652
C1=CC(=CC=C1CC(C(=O)O)N)Cl
C9H10ClNO2
MFCD00002601
fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro
2-amino-3-(4-chlorophenyl)propanoic acid

Specifications

Authentic
0.5% max. (K.F.)
5g
98%
ClC6H4CH2CH(NH2)CO2H
14, I, 608
NIGWMJHCCYYCSF-UHFFFAOYSA-N
2-amino-3-(4-chlorophenyl)propanoic acid
4652
Powder
DL-4-Chlorophenylalanine, 98%
Glass bottle
White
7424-00-2
C9H10ClNO2
MFCD00002601
fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro
C1=CC(=CC=C1CC(C(=O)O)N)Cl
199.634
199.64
≥97.5%
Safety and Handling

Safety and Handling

EINECSNumber : 231-051-9

RTECSNumber : AY4390010

TSCA : TSCA