Please login to your online account to display your discounted pricing

DL-4-Amino-3-hydroxybutyric Acid 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103810250

 View more versions of this product

Catalog No. AC103810250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 119.12
Physical Form Crystalline Powder
Quantity 25g
Color White
TSCA TSCA
Assay 97.5 to 102.5%
EINECS Number 213-106-9
Chemical Name or Material DL-4-Amino-3-hydroxybutyric acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4H9NO3
Linear Formula H2NCH2CH(OH)CH2CO2H
MDL Number MFCD00008141
CAS 924-49-2
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material DL-4-Amino-3-hydroxybutyric acid
Name Note 98%
CAS 924-49-2
Assay 97.5 to 102.5%
Assay Percent Range 98%
Molecular Formula C4H9NO3
Linear Formula H2NCH2CH(OH)CH2CO2H
Formula Weight 119.12
MDL Number MFCD00008141