Please login to your online account to display your discounted pricing

DL-4-Amino-3-hydroxybutyric Acid 98%, ACROS Organics™

$125.43 - $559.69

Chemical Identifiers

Chemical Name or Material DL-4-Amino-3-hydroxybutyric acid
Name Note 98%
CAS 924-49-2
Assay 97.5 to 102.5%
Assay Percent Range 98%
Molecular Formula C4H9NO3
Linear Formula H2NCH2CH(OH)CH2CO2H
Formula Weight 119.12
MDL Number MFCD00008141
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC103810050

  SDS
thermo fisher scientific
103810050
Glass bottle 5g Each for $125.43

AC103810250

  SDS
thermo fisher scientific
103810250
Glass bottle 25g Each for $559.69
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 119.12
Quantity 5g
Color White
Assay 97.5 to 102.5%
TSCA TSCA
EINECS Number 213-106-9
CAS 924-49-2
Molecular Formula C4H9NO3
Assay Percent Range 98%
Name Note 98%
Chemical Name or Material DL-4-Amino-3-hydroxybutyric acid
MDL Number MFCD00008141
Linear Formula H2NCH2CH(OH)CH2CO2H
Packaging Glass bottle
Infrared Spectrum Authentic