missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-3-Phenyllactic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 427770100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
DL-3-Phenyllactic acid | |
Authentic | |
Glass bottle | |
C6H5CH2CH(OH)COOH | |
10g | |
VOXXWSYKYCBWHO-UHFFFAOYSA-N | |
2-hydroxy-3-phenylpropanoic acid | |
3848 | |
166.18 |
828-01-3 | |
96% min. (HPLC) | |
C9H10O3 | |
MFCD00065928 | |
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid | |
C1=CC=C(C=C1)CC(C(=O)O)O | |
166.18 | |
CHEBI:25998 | |
97% |
Chemical Identifiers
828-01-3 | |
166.18 | |
VOXXWSYKYCBWHO-UHFFFAOYSA-N | |
3848 | |
2-hydroxy-3-phenylpropanoic acid |
C9H10O3 | |
MFCD00065928 | |
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid | |
CHEBI:25998 | |
C1=CC=C(C=C1)CC(C(=O)O)O |
Safety and Handling
EINECSNumber : 212-580-4
RUO â Research Use Only