missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-3-Octanol, 97%, ACROS Organics™

Manufacturer:  Acros Organics 203770500

 View more versions of this product

Catalog No. AC203770500



Specifications

Specifications

DL-3-Octanol
67°C
Glass bottle
0.825
Colorless
50mL
97%
CH3(CH2)4CH(OH)C2H5
01, I, 208
Solubility in water: insoluble.
CCCCCC(CC)O
130.23
CHEBI:80945
Liquid
0.8250g/mL
Authentic
1.4220 to 1.4280
174.0°C to 176.0°C
-45.0°C
589-98-0
C8H18O
MFCD00004590
1-ethylhexanol
NMRPBPVERJPACX-UHFFFAOYSA-N
octan-3-ol
11527
130.23
97%

Chemical Identifiers

589-98-0
130.23
NMRPBPVERJPACX-UHFFFAOYSA-N
11527
octan-3-ol
C8H18O
MFCD00004590
1-ethylhexanol
CHEBI:80945
CCCCCC(CC)O
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents