Please login to your online account to display your discounted pricing

DL-3-(Methylthio)-2-butanone, 99%, Acros Organics


Quantity 100g

Manufacturer: thermo fisher scientific  208981000

 View more versions of this product

Catalog No. AC208981000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear yellow to light greenish
Formula Weight 118.19
Assay ≥98.5%
Physical Form Liquid
Quantity 100g
Beilstein 01, IV, 3993
EINECS Number 258-576-6
Density 0.97g/mL
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.4700 to 1.4720 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material DL-3-(Methylthio)-2-butanone
MDL Number MFCD00008761
Molecular Formula C5H10OS
Name Note 99%
CAS 53475-15-3
Assay Percent Range 99%
Linear Formula CH3CH(SCH3)COCH3

Specifications

Chemical Name or Material DL-3-(Methylthio)-2-butanone
Name Note 99%
CAS 53475-15-3
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H10OS
Linear Formula CH3CH(SCH3)COCH3
Formula Weight 118.19
MDL Number MFCD00008761
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.
Highly flammable liquid and vapor.

SDS & Safety Documents