Please login to your online account to display your discounted pricing

DL-3-Hydroxybutyric acid, sodium salt, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC215011000

 View more versions of this product

Catalog No. AC215011000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 126.09
Melting Point 161° to 167°C (dec.)
Quantity 100g
Color White
Beilstein 03, 308
Chemical Name or Material DL-3-Hydroxybutyric acid, sodium salt
Linear Formula CH3CH(OH)CH2CO2Na
Assay Percent Range 99%
Molecular Formula C4H7NaO3
MDL Number MFCD00016716
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 150-83-4

Specifications

Chemical Name or Material DL-3-Hydroxybutyric acid, sodium salt
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H7NaO3
Linear Formula CH3CH(OH)CH2CO2Na
Formula Weight 126.09
MDL Number MFCD00016716