missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-3-Hydroxybutyric acid, sodium salt, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 215010250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-3-Hydroxybutyric acid, sodium salt | |
161.0°C to 167.0°C | |
Authentic | |
Glass bottle | |
CH3CH(OH)CH2CO2Na | |
25 g | |
sodium 3-hydroxybutyrate, sodium 3-hydroxybutanoate, dl-3-hydroxybutyric acid sodium salt, sodium beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, monosodium salt, butanoic acid,3-hydroxy-, sodium salt 1:1, dl-3-hydroxybutyric acid, sodium salt, 3-hydroxybutyric acid sodium salt, sodium 3-oxidanylbutanoate, dl-beta-hydroxybutyric acid sodium salt | |
NBPUSGBJDWCHKC-UHFFFAOYSA-M | |
sodium;3-hydroxybutanoate | |
23676771 | |
99% |
150-83-4 | |
White | |
99% | |
C4H7NaO3 | |
MFCD00016716 | |
03, 308 | |
Solubility in water: freely soluble | |
CC(CC(=O)[O-])O.[Na+] | |
126.09 | |
126.09 | |
Crystalline Powder |
Chemical Identifiers
150-83-4 | |
126.09 | |
NBPUSGBJDWCHKC-UHFFFAOYSA-M | |
23676771 | |
CC(CC(=O)[O-])O.[Na+] |
C4H7NaO3 | |
MFCD00016716 | |
sodium 3-hydroxybutyrate, sodium 3-hydroxybutanoate, dl-3-hydroxybutyric acid sodium salt, sodium beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, monosodium salt, butanoic acid,3-hydroxy-, sodium salt 1:1, dl-3-hydroxybutyric acid, sodium salt, 3-hydroxybutyric acid sodium salt, sodium 3-oxidanylbutanoate, dl-beta-hydroxybutyric acid sodium salt | |
sodium;3-hydroxybutanoate |
Safety and Handling
EINECSNumber : 205-774-5
RUO – Research Use Only