Please login to your online account to display your discounted pricing

DL-3-Amino-n-butyric acid, 97%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  103270250

 View more versions of this product

Catalog No. AC103270250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 103.12
Physical Form Powder
Quantity 25g
Color White
Beilstein 04, 412
Merck Index 15, 424
Chemical Name or Material DL-3-Amino-n-butyric acid
Water 1% max.
Assay Percent Range 97%
Molecular Formula C4H9NO2
Linear Formula CH3CH(NH2)CH2CO2H
CAS 541-48-0
Infrared Spectrum Authentic

Specifications

Chemical Name or Material DL-3-Amino-n-butyric acid
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H9NO2
Linear Formula CH3CH(NH2)CH2CO2H
Formula Weight 103.12