missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-3-Amino-n-butyric acid, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C4H9NO2
Linear Formula CH3CH(NH2)CH2CO2H
Formula Weight 103.12
Chemical Name or Material DL-3-Amino-n-butyric acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC103270250 SDS Thermo Fisher Scientific
103270250
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
CH3CH(NH2)CH2CO2H
DL-3-Amino-n-butyric acid
C4H9NO2
103.12
≥96.0%

Specifications

103.12
25g
04, 412
1% max.
541-48-0
CH3CH(NH2)CH2CO2H
DL-3-Amino-n-butyric acid
Powder
White
15, 424
97%
C4H9NO2
Authentic
≥96.0%