Please login to your online account to display your discounted pricing

DL-3-Amino-3-phenylpropionic acid, 97%, ACROS Organics™

$416.54 - $416.54

Chemical Identifiers

Chemical Name or Material DL-3-Amino-3-phenylpropionic acid
Name Note tech., 90%
CAS 614-19-7
Purity Grade Technical Grade
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H11NO2
Linear Formula C6H5CH(NH2)CH2CO2H
Formula Weight 165.19
MDL Number MFCD00008064
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170380250

  SDS
thermo fisher scientific
170380250
Glass bottle 25g Each for $416.54
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 165.19
Physical Form Powder
Purity Grade Technical Grade
Quantity 25g
Color White to slightly yellow
Beilstein 14, 493
EINECS Number 210-371-2
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Molecular Formula C9H11NO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 614-19-7
Linear Formula C6H5CH(NH2)CH2CO2H
Assay Percent Range 97%
MDL Number MFCD00008064
Chemical Name or Material DL-3-Amino-3-phenylpropionic acid
Name Note tech., 90%
Synonym β-Aminohydrocinnamic acid
Infrared Spectrum Authentic
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.