missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Pyrrolidone-5-carboxylic acid, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 232930050

 View more versions of this product

Catalog No. AC232930050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Crystalline Powder
White
20ppm max.
0.5% max.
MFCD00064322
DL-2-Pyrrolidone-5-carboxylic acid
98%
0.3% max.
200ppm max.
DL-Pyroglutamic acid
129.12
5g
300ppm max.
700ppm max.
C5H7NO3
149-87-1
Glass bottle
30ppm max.
2ppm max.
Authentic

Chemical Identifiers

149-87-1
C5H7NO3
MFCD00064322
DL-2-Pyrrolidone-5-carboxylic acid
98%
129.12
DL-Pyroglutamic acid
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.