missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-2-Phenylglycinol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 374280050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-2-Phenylglycinol | |
53.0°C to 58.0°C | |
261.0°C | |
97.5% min. (GC) | |
C8H11NO | |
1.5484 | |
5 g | |
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino | |
NC(CO)C1=CC=CC=C1 | |
137.18 | |
137.18 | |
Powder |
7568-92-5 | |
White to Yellow | |
Authentic | |
Glass bottle | |
C6H5CH(NH2)CH2OH | |
MFCD00130145 | |
13,IV,1835 | |
IJXJGQCXFSSHNL-UHFFFAOYNA-N | |
2-amino-2-phenylethan-1-ol | |
92466 | |
98% |
Chemical Identifiers
7568-92-5 | |
137.18 | |
IJXJGQCXFSSHNL-UHFFFAOYNA-N | |
92466 |
C8H11NO | |
MFCD00130145 | |
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino | |
NC(CO)C1=CC=CC=C1 |
Safety and Handling
GHS Signal Word: Danger
RUO – Research Use Only