Please login to your online account to display your discounted pricing

DL-2-Phenylglycinol, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC374280250

 View more versions of this product

Catalog No. AC374280250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.5% (GC)
Formula Weight 137.18
Quantity 25g
Color White to light yellow
Assay Percent Range 98%
Linear Formula C6H5CH(NH2)CH2OH
Chemical Name or Material DL-2-Phenylglycinol
Packaging Glass bottle
Molecular Formula C8H11NO
CAS 7568-92-5
Synonym beta-Aminophenethyl alcohol
Infrared Spectrum Authentic

Specifications

Chemical Name or Material DL-2-Phenylglycinol
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C8H11NO
Linear Formula C6H5CH(NH2)CH2OH
Formula Weight 137.18
Synonym beta-Aminophenethyl alcohol