missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-2-Phenylglycine, 98%

$25.12 - $172.03

Chemical Identifiers

CAS 2835-06-5
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00064402
Beilstein 3197862
Merck Index 14,7291
Synonym (+/-)-alpha-Aminophenylacetic acid; H-DL-Phg-OH
Chemical Name or Material DL-2-Phenylglycine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2112922 SDS Alfa Aesar™
B2112922
100g Each for $43.82
AAB2112936 SDS Alfa Aesar™
B2112936
500g Each for $172.03
AAB2112914 SDS Alfa Aesar™
B2112914
25g Each for $25.12
Specifications Description & Specifications

Chemical Identifiers

2835-06-5
151.17
3197862
(+/-)-alpha-Aminophenylacetic acid; H-DL-Phg-OH
C8H9NO2
MFCD00064402
14,7291
DL-2-Phenylglycine

Specifications

2835-06-5
151.17
3197862
(+/-)-alpha-Aminophenylacetic acid; H-DL-Phg-OH
100g
DL-2-Phenylglycine
C8H9NO2
MFCD00064402
14,7291
≈290°C (subl.)
Odorless
98%

Safety and Handling

TSCA : TSCA