missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Phenylglycinamide, TCI America™

Manufacturer:  TCI America P24025G

 View more versions of this product

Catalog No. P24025G



Specifications

Specifications

DL-2-Phenylglycinamide
130°C
700-63-0
C8H10N2O
2-Amino-2-phenylacetamide
C1=CC=C(C=C1)C(C(=O)N)N
150.181
150.18
White-Yellow
5g
≥98.0% (HPLC)
MFCD00025498
KIYRSYYOVDHSPG-UHFFFAOYSA-N
2-amino-2-phenylacetamide
12791
Crystal-Powder at 20°C

Chemical Identifiers

700-63-0
150.181
KIYRSYYOVDHSPG-UHFFFAOYSA-N
12791
C1=CC=C(C=C1)C(C(=O)N)N
C8H10N2O
MFCD00025498
2-Amino-2-phenylacetamide
2-amino-2-phenylacetamide
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0084

RTECSNumber : AB4390000

TSCA : No

Documents