missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Methylglutamic Acid Hemihydrate 98.0+%, TCI America™

$46.08 - $46.08

Chemical Identifiers

CAS 71-90-9
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.157
MDL Number MFCD00004188
InChI Key QHSCIWIRXWFIGH-UHFFFAOYSA-N
Synonym 2-amino-2-methylglutaric acid, 2-methylglutamic acid, 2-methylglutamic acid, dl, a-methylglutamic acid, alpha-methylglutamate, alpha-methylglutamic acid, dl-2-methylglutamic acid, dl-glutamic acid, 2-methyl, glutamic acid, 2-methyl, hemi-hydrate
PubChem CID 95440
IUPAC Name 2-amino-2-methylpentanedioic acid
SMILES CC(CCC(=O)O)(C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M0229100MG View Documents Tci America
M0229100MG
100mg Each for $46.08
 
Specifications

Chemical Identifiers

71-90-9
161.157
QHSCIWIRXWFIGH-UHFFFAOYSA-N
95440
CC(CCC(=O)O)(C(=O)O)N
C6H11NO4
MFCD00004188
2-amino-2-methylglutaric acid, 2-methylglutamic acid, 2-methylglutamic acid, dl, a-methylglutamic acid, alpha-methylglutamate, alpha-methylglutamic acid, dl-2-methylglutamic acid, dl-glutamic acid, 2-methyl, glutamic acid, 2-methyl, hemi-hydrate
2-amino-2-methylpentanedioic acid

Specifications

White
100mg
C6H11NO4
2-amino-2-methylglutaric acid, 2-methylglutamic acid, 2-methylglutamic acid, dl, a-methylglutamic acid, alpha-methylglutamate, alpha-methylglutamic acid, dl-2-methylglutamic acid, dl-glutamic acid, 2-methyl, glutamic acid, 2-methyl, hemi-hydrate
CC(CCC(=O)O)(C(=O)O)N
161.157
161.16
≥98.0% (T)
160°C
71-90-9
MFCD00004188
QHSCIWIRXWFIGH-UHFFFAOYSA-N
2-amino-2-methylpentanedioic acid
95440
Crystal-Powder at 20°C
DL-2-Methylglutamic Acid Hemihydrate
Safety and Handling

Safety and Handling

TSCA : No