missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Methylbutyryl chloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 414151000

 View more versions of this product

Catalog No. AC414151000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

DL-2-Methylbutyryl chloride
0.98g/mL
Glass bottle
1.4160 to 1.4180 (20°C, 589nm)
98%
CH3CH2CH(CH3)COCl
MFCD00041726
100g
57526-28-0
Authentic
Liquid
≥97.5% (RCOCL)
C5H9ClO
120.58
Clear colorless to light yellow

Chemical Identifiers

98%
CH3CH2CH(CH3)COCl
MFCD00041726
≥97.5% (RCOCL)
C5H9ClO
120.58
DL-2-Methylbutyryl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger