Please login to your online account to display your discounted pricing

DL-2-Methylbutyryl chloride, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC414150010

 View more versions of this product

Catalog No. AC414150010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (RCOCL)
Formula Weight 120.58
Physical Form Liquid
Quantity 1g
Color Clear colorless to light yellow
Density 0.98g/mL
Refractive Index 1.4160 to 1.4180 (20°C, 589nm)
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)COCl
CAS 57526-28-0
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
MDL Number MFCD00041726
Infrared Spectrum Authentic
Molecular Formula C5H9ClO
Chemical Name or Material DL-2-Methylbutyryl chloride
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material DL-2-Methylbutyryl chloride
Assay ≥97.5% (RCOCL)
Assay Percent Range 98%
Molecular Formula C5H9ClO
Linear Formula CH3CH2CH(CH3)COCl
Formula Weight 120.58
MDL Number MFCD00041726
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents