missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-2-Methylbutyric Acid 97.0+%, TCI America™
Supplier: TCI America M0181500ML
Specifications
DL-2-Methylbutyric Acid | |
Yellow | |
C5H10O2 | |
500 mL | |
2-methylbutyric acid, dl-2-methylbutyric acid, butanoic acid, 2-methyl, ethylmethylacetic acid, methylbutyric acid, 2-methyl butyric acid, 2-methybutyric acid, methylethylacetic acid, active valeric acid, butyric acid, 2-methyl | |
CCC(C)C(=O)O | |
102.133 | |
CHEBI:37070 | |
≥97.0% (GC) |
116-53-0 | |
174°C | |
MFCD00002669 | |
3265 | |
WLAMNBDJUVNPJU-UHFFFAOYSA-N | |
2-methylbutanoic acid | |
8314 | |
102.13 | |
Liquid |
Chemical Identifiers
116-53-0 | |
102.133 | |
WLAMNBDJUVNPJU-UHFFFAOYSA-N | |
8314 | |
2-methylbutanoic acid |
C5H10O2 | |
MFCD00002669 | |
2-methylbutyric acid, dl-2-methylbutyric acid, butanoic acid, 2-methyl, ethylmethylacetic acid, methylbutyric acid, 2-methyl butyric acid, 2-methybutyric acid, methylethylacetic acid, active valeric acid, butyric acid, 2-methyl | |
CHEBI:37070 | |
CCC(C)C(=O)O |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : EK7897000
TSCA : Yes