Please login to your online account to display your discounted pricing

DL-2-Isopropylserine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material DL-2-Isopropylserine
Name Note 99%
CAS 7522-43-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H13NO3
Formula Weight 147.17
MDL Number MFCD01863303
Synonym DL-2-Amino-2-hydroxymethyl-3-methylbutyric acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC339760010

  SDS
thermo fisher scientific
339760010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC339760050

  SDS
thermo fisher scientific
339760050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 147.17
Quantity 1g
Color Cream
Chemical Name or Material DL-2-Isopropylserine
Name Note 99%
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C6H13NO3
MDL Number MFCD01863303
CAS 7522-43-2
Synonym DL-2-Amino-2-hydroxymethyl-3-methylbutyric acid
Packaging Glass bottle