Please login to your online account to display your discounted pricing

DL-2-Bromopropionic acid, 99+%, ACROS Organics™


Quantity 2.5kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC107350025

 View more versions of this product

Catalog No. AC107350025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid After Melting
Assay ≥99.0%
Formula Weight 152.98
Quantity 2.5kg
Color Clear colorless to yellow
Density 1.6500g/mL
Beilstein 02, 254
Chemical Name or Material DL-2-Bromopropionic acid
Assay Percent Range 99+%
Molecular Formula C3H5BrO2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4740 to 1.4760 (20°C, 589nm)
Linear Formula CH3CH(Br)CO2H
MDL Number MFCD00004211
CAS 598-72-1
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Packaging Glass bottle

Specifications

Chemical Name or Material DL-2-Bromopropionic acid
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C3H5BrO2
Linear Formula CH3CH(Br)CO2H
Formula Weight 152.98
MDL Number MFCD00004211
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents