missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Bromopropionic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107352500

 View more versions of this product

Catalog No. AC107352500


 

Specifications Description & Specifications

Specifications

Liquid After Melting
152.98
Clear colorless to yellow
CH3CH(Br)CO2H
Glass bottle
1.4740 to 1.4760 (20°C, 589nm)
99+%
MFCD00004211
≥99.0%
250g
02, 254
Authentic
598-72-1
C3H5BrO2
DL-2-Bromopropionic acid

Chemical Identifiers

99+%
CH3CH(Br)CO2H
MFCD00004211
≥99.0%
C3H5BrO2
152.98
DL-2-Bromopropionic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger