Learn More
DL-2-Benzylserine, 97%, ACROS Organics™
$97.63 - $97.63
Chemical Identifiers
CAS | 4740-47-0 |
---|---|
Molecular Formula | C10H13NO3 |
Molecular Weight (g/mol) | 195.22 |
MDL Number | MFCD01863305 |
InChI Key | ZMNNAJIBOJDHAF-JTQLQIEISA-N |
Synonym | 2-amino-3-hydroxy-2-benzylpropanoic acid, alpha-benzyl-dl-serine, dl-2-benzylserine |
PubChem CID | 7010088 |
IUPAC Name | (2S)-2-amino-2-benzyl-3-hydroxypropanoic acid |
SMILES | C1=CC=C(C=C1)CC(CO)(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC339780010
|
Thermo Fisher Scientific
339780010 |
1g | Glass bottle |
Each for $97.63
|
|
AC339780100
|
Thermo Fisher Scientific
AC339780100 |
10g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
4740-47-0 | |
195.22 | |
ZMNNAJIBOJDHAF-JTQLQIEISA-N | |
7010088 | |
C1=CC=C(C=C1)CC(CO)(C(=O)O)N |
C10H13NO3 | |
MFCD01863305 | |
2-amino-3-hydroxy-2-benzylpropanoic acid, alpha-benzyl-dl-serine, dl-2-benzylserine | |
(2S)-2-amino-2-benzyl-3-hydroxypropanoic acid |
Specifications
Authentic | |
Beige to Brown | |
1g | |
97% | |
MFCD01863305 | |
Solubility in water: soluble in water | |
C1=CC=C(C=C1)CC(CO)(C(=O)O)N | |
195.22 | |
195.22 | |
97% |
Glass bottle | |
245°C to 263°C | |
4740-47-0 | |
C10H13NO3 | |
2-amino-3-hydroxy-2-benzylpropanoic acid, alpha-benzyl-dl-serine, dl-2-benzylserine | |
ZMNNAJIBOJDHAF-JTQLQIEISA-N | |
(2S)-2-amino-2-benzyl-3-hydroxypropanoic acid | |
7010088 | |
Powder or Flakes | |
DL-2-Benzylserine, 97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning