missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Aminoadipic acid hydrate, 97%, ACROS Organics™

Chemical Identifiers

CAS 542-32-5,na
Assay Percent Range 97%
Molecular Formula C6H11NO4·xH2O
Linear Formula HO2C(CH2)3CH(NH2)CO2H·xH2O
Formula Weight 161.16
MDL Number MFCD00150708
Chemical Name or Material DL-2-Aminoadipic acid hydrate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC154500050 SDS Thermo Fisher Scientific
154500050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

542-32-5,na
C6H11NO4·xH2O
161.16
DL-2-Aminoadipic acid hydrate
97%
HO2C(CH2)3CH(NH2)CO2H·xH2O
MFCD00150708
≥96.0%

Specifications

161.16
5g
14, 416
Authentic
542-32-5,na
C6H11NO4·xH2O
Glass bottle
≥96.0%
Powder
White
04, 495
HO2C(CH2)3CH(NH2)CO2H·xH2O
97%
MFCD00150708
DL-2-Aminoadipic acid hydrate