Please login to your online account to display your discounted pricing

DL-2-Aminoadipic acid hydrate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  154500050

 View more versions of this product

Catalog No. AC154500050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.16
Assay ≥96.0%
Physical Form Powder
Quantity 5g
Color White
Merck Index 14, 416
Beilstein 04, 495
Infrared Spectrum Authentic
Linear Formula HO2C(CH2)3CH(NH2)CO2H·xH2O
Chemical Name or Material DL-2-Aminoadipic acid hydrate
CAS 542-32-5
Assay Percent Range 97%
Molecular Formula C6H11NO4·xH2O
MDL Number MFCD00150708
Packaging Glass bottle
CAS na

Specifications

Chemical Name or Material DL-2-Aminoadipic acid hydrate
CAS 542-32-5
CAS na
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H11NO4·xH2O
Linear Formula HO2C(CH2)3CH(NH2)CO2H·xH2O
Formula Weight 161.16
MDL Number MFCD00150708