missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-2-Aminoadipic acid, 99%, anhydrous, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441790010
Specifications
DL-2-Aminoadipic acid | |
Glass bottle | |
176.0°C to 177.0°C | |
542-32-5 | |
C6H11NO4 | |
2-amino-hexanedioic acid, 2-aminoadipate, 2-aminoadipic acid, alpha-aminoadipate, alpha-aminoadipic acid, aminoadipic acid, dl-2-aminoadipic acid, dl-2-aminohexanedioic acid, dl-alpha-aminoadipic acid, hexanedioic acid, 2-amino | |
OYIFNHCXNCRBQI-UHFFFAOYSA-N | |
2-aminohexanedioic acid | |
469 | |
161.16 | |
99% |
Authentic | |
White | |
1g | |
99% | |
15,41 | |
Solubility in water: .. Other solubilities: sparingly soluble in alcohol and ether | |
C(CC(C(=O)O)N)CC(=O)O | |
161.16 | |
CHEBI:37024 | |
Powder or Crystalline Powder |
Chemical Identifiers
542-32-5 | |
161.16 | |
2-amino-hexanedioic acid, 2-aminoadipate, 2-aminoadipic acid, alpha-aminoadipate, alpha-aminoadipic acid, aminoadipic acid, dl-2-aminoadipic acid, dl-2-aminohexanedioic acid, dl-alpha-aminoadipic acid, hexanedioic acid, 2-amino | |
CHEBI:37024 | |
C(CC(C(=O)O)N)CC(=O)O |
C6H11NO4 | |
OYIFNHCXNCRBQI-UHFFFAOYSA-N | |
469 | |
2-aminohexanedioic acid |
Safety and Handling
EINECSNumber : 208-809-2