Please login to your online account to display your discounted pricing

DL-2-Aminoadipic acid, 99%, anhydrous, ACROS Organics™

$79.49 - $264.55

Chemical Identifiers

Chemical Name or Material DL-2-Aminoadipic acid
Assay 98.5 to 101.5%
Assay Percent Range 99%
Molecular Formula C6H11NO4
Formula Weight 161.16
Synonym (R, S)-2-Aminohexanedioic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441790010

 
thermo fisher scientific
AC441790010
1g Each for $79.49

AC441790050

 
thermo fisher scientific
AC441790050
5g Each for $264.55
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Formula Weight 161.16
Quantity 1g
Color White to off-white
Assay 98.5 to 101.5%
Molecular Formula C6H11NO4
CAS 542-32-5
Assay Percent Range 99%
Chemical Name or Material DL-2-Aminoadipic acid
Synonym (R, S)-2-Aminohexanedioic acid
Infrared Spectrum Authentic