missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-2-Amino-4-phosphonobutyric acid, 95%

$212.81 - $609.14

Chemical Identifiers

CAS 20263-07-4
Molecular Formula C4H10NO5P
Molecular Weight (g/mol) 183.1
MDL Number MFCD00013999
InChI Key DDOQBQRIEWHWBT-UHFFFAOYSA-N
Synonym 2-amino-4-phosphonobutyric acid, dl-ap4, dl-2-amino-4-phosphonobutyric acid, butanoic acid, 2-amino-4-phosphono, +/--2-amino-4-phosphonobutyric acid, butyric acid, 2-amino-4-phosphono, 2-amino-4-phosphono-butyric acid, l-2-amino-4-phosphonobutyrate, l-apb, abu p
PubChem CID 2207
IUPAC Name 2-amino-4-phosphonobutanoic acid
SMILES C(CP(=O)(O)O)C(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1767103 View Documents Alfa Aesar™
L1767103
1g Each for $609.14
 
AAL17671MD View Documents Alfa Aesar™
L17671MD
250mg Each for $212.81
 
Specifications

Chemical Identifiers

20263-07-4
183.1
DDOQBQRIEWHWBT-UHFFFAOYSA-N
2207
C(CP(=O)(O)O)C(C(=O)O)N
C4H10NO5P
MFCD00013999
2-amino-4-phosphonobutyric acid, dl-ap4, dl-2-amino-4-phosphonobutyric acid, butanoic acid, 2-amino-4-phosphono, +/--2-amino-4-phosphonobutyric acid, butyric acid, 2-amino-4-phosphono, 2-amino-4-phosphono-butyric acid, l-2-amino-4-phosphonobutyrate, l-apb, abu p
2-amino-4-phosphonobutanoic acid

Specifications

225°C to 228°C
20263-07-4
MFCD00013999
2-amino-4-phosphonobutyric acid, dl-ap4, dl-2-amino-4-phosphonobutyric acid, butanoic acid, 2-amino-4-phosphono, +/--2-amino-4-phosphonobutyric acid, butyric acid, 2-amino-4-phosphono, 2-amino-4-phosphono-butyric acid, l-2-amino-4-phosphonobutyrate, l-apb, abu p
C(CP(=O)(O)O)C(C(=O)O)N
183.1
183.1
DL-2-Amino-4-phosphonobutyric acid
1g
C4H10NO5P
2093908
DDOQBQRIEWHWBT-UHFFFAOYSA-N
2-amino-4-phosphonobutanoic acid
2207
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning