Please login to your online account to display your discounted pricing

DL-2-Amino-3-methyl-1-butanol, 95%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  180420010

 View more versions of this product

Catalog No. AC180420010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 103.16
Quantity 1g
Color Clear yellow
EINECS Number 240-425-0
Chemical Name or Material DL-2-Amino-3-methyl-1-butanol
Refractive Index 1.4533 to 1.4553 (20°C, 589nm)
Name Note 95%
Assay Percent Range 95%
Molecular Formula C5H13NO
Linear Formula (CH3)2CHCH(NH2)CH2OH
MDL Number MFCD00004730
CAS 16369-05-4
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Synonym DL-Valinol
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material DL-2-Amino-3-methyl-1-butanol
Name Note 95%
CAS 16369-05-4
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H13NO
Linear Formula (CH3)2CHCH(NH2)CH2OH
Formula Weight 103.16
MDL Number MFCD00004730
Synonym DL-Valinol
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents