Please login to your online account to display your discounted pricing

DL-2-Amino-1-propanol, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  185080250

 View more versions of this product

Catalog No. AC185080250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Viscous Liquid
Formula Weight 75.11
Quantity 25g
Color Clear colorless to light yellow
Merck Index 10, 472
Beilstein 04, I, 432
EINECS Number 228-207-3
TSCA TSCA
Specific Rotation -0.5° to +0.5° (20°C, 589nm) (neat)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C3H9NO
MDL Number MFCD00008085
Name Note 98%
CAS 6168-72-5
Chemical Name or Material DL-2-Amino-1-propanol
Assay Percent Range 98%
Linear Formula CH3CH(NH2)CH2OH
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4480 to 1.4510 (20°C, 589nm)
Synonym DL-Alaninol
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material DL-2-Amino-1-propanol
Name Note 98%
CAS 6168-72-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H9NO
Linear Formula CH3CH(NH2)CH2OH
Formula Weight 75.11
MDL Number MFCD00008085
Synonym DL-Alaninol
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents