Please login to your online account to display your discounted pricing

DL-2-Amino-1-propanol, 98%, ACROS Organics™

$74.12 - $245.27

Chemical Identifiers

Chemical Name or Material DL-2-Amino-1-propanol
Name Note 98%
CAS 6168-72-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H9NO
Linear Formula CH3CH(NH2)CH2OH
Formula Weight 75.11
MDL Number MFCD00008085
Synonym DL-Alaninol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC185080050

  SDS
thermo fisher scientific
185080050
Glass bottle 5g Each for $74.12

AC185080250

  SDS
thermo fisher scientific
185080250
Glass bottle 25g Each for $245.27
Description & Specifications

Specifications

Physical Form Viscous Liquid
Assay ≥97.5%
Formula Weight 75.11
Quantity 5g
Color Clear colorless to light yellow
EINECS Number 228-207-3
TSCA TSCA
Beilstein 04, I, 432
Merck Index 10, 472
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4480 to 1.4510 (20°C, 589nm)
Chemical Name or Material DL-2-Amino-1-propanol
Name Note 98%
Assay Percent Range 98%
Molecular Formula C3H9NO
Linear Formula CH3CH(NH2)CH2OH
CAS 6168-72-5
Specific Rotation -0.5° to +0.5° (20°C, 589nm) (neat)
MDL Number MFCD00008085
Infrared Spectrum Authentic
Synonym DL-Alaninol
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.