Please login to your online account to display your discounted pricing

DL-2,3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  186740010

 View more versions of this product

Catalog No. AC186740010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 228.28
Quantity 1g
Color White to practically white
Merck Index 14, 3210
EINECS Number 223-796-3
RTECS Number TZ6420000
Loss on Drying 7% to 9% (1 g, 100°C)
Synonym DMPS sodium salt monohydrate
Health Hazard 1 AIR SENSITIVE
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material DL-2, 3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate
MDL Number MFCD00149543
Assay Percent Range 95%
CAS 207233-91-8
Linear Formula HSCH2CH(SH)CH2SO3Na·H2O
Molecular Formula C3H7NaO3S3·H2O
Name Note 95%

Specifications

Chemical Name or Material DL-2, 3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate
Name Note 95%
CAS 207233-91-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C3H7NaO3S3·H2O
Linear Formula HSCH2CH(SH)CH2SO3Na·H2O
Formula Weight 228.28
MDL Number MFCD00149543
Synonym DMPS sodium salt monohydrate
Accessories

AIR SENSITIVE

SDS & Safety Documents