Please login to your online account to display your discounted pricing

DL-2,3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material DL-2, 3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate
Name Note 95%
CAS 207233-91-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C3H7NaO3S3·H2O
Linear Formula HSCH2CH(SH)CH2SO3Na·H2O
Formula Weight 228.28
MDL Number MFCD00149543
Synonym DMPS sodium salt monohydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC186740010

  SDS
fisher scientific acros
186740010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC186742500

  SDS
fisher scientific acros
186742500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 228.28
Quantity 1g
Color White to practically white
Merck Index 14, 3210
EINECS Number 223-796-3
RTECS Number TZ6420000
Loss on Drying 7% to 9% (1 g, 100°C)
Synonym DMPS sodium salt monohydrate
Health Hazard 1 AIR SENSITIVE
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material DL-2, 3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate
MDL Number MFCD00149543
Assay Percent Range 95%
CAS 207233-91-8
Linear Formula HSCH2CH(SH)CH2SO3Na·H2O
Molecular Formula C3H7NaO3S3·H2O
Name Note 95%
Safety and Handling

AIR SENSITIVE