Please login to your online account to display your discounted pricing

dl-1-Amino-2-propanol ≥99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  149650250

 View more versions of this product

Catalog No. AC149650250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥93%
Formula Weight 75.11
Physical Form Liquid
Quantity 25g
Color Clear
TSCA TSCA
RTECS Number UA5775000
EINECS Number 201-162-7
Beilstein 04, 289
Infrared Spectrum Authentic
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 94%
Chemical Name or Material DL-1-Amino-2-propanol
Linear Formula CH3CH(OH)CH2NH2
Molecular Formula C3H9NO
MDL Number MFCD00008139
CAS 78-96-6
Name Note 99+%
Packaging Glass bottle
Synonym DL-Isopropanolamine
Water 0.5% max. (K.F.)
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material DL-1-Amino-2-propanol
Name Note 99+%
CAS 78-96-6
Assay ≥93%
Assay Percent Range 94%
Molecular Formula C3H9NO
Linear Formula CH3CH(OH)CH2NH2
Formula Weight 75.11
MDL Number MFCD00008139
Synonym DL-Isopropanolamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents