missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-1-(2-Furyl)ethanol, 99%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 339670010

 View more versions of this product

Catalog No. AC339670010


 

Specifications Description & Specifications

Specifications

DL-1-(2-Furyl)ethanol
4208-64-4
Liquid
99%
C6H8O2
112.13
17, V,3, 374
DL-α-Methylfuran-2-methanol
1.0700g/mL
1g
99%
Authentic
≥98.5%
Glass bottle
1.4770 to 1.4800 (20°C, 589nm)
MFCD00077775
0.3 to 0.5% hydroquinone
Product may darken on storage
Clear colorless to light yellow

Chemical Identifiers

4208-64-4
C6H8O2
MFCD00077775
DL-1-(2-Furyl)ethanol, 99%
99%
112.13
DL-α-Methylfuran-2-methanol
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 224-130-4

TSCA : TSCA