Please login to your online account to display your discounted pricing

DL-1-(2-Furyl)ethanol, 99%, stabilized, ACROS Organics™

$56.81 - $56.81

Chemical Identifiers

Chemical Name or Material DL-1-(2-Furyl)ethanol
Name Note 99%
CAS 4208-64-4
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H8O2
Formula Weight 112.13
MDL Number MFCD00077775
Synonym DL-α-Methylfuran-2-methanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC339670010

  SDS
thermo fisher scientific
339670010
Glass bottle 1g Each for $56.81

AC339670050

  SDS
thermo fisher scientific
339670050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 112.13
Quantity 1g
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 224-130-4
Density 1.0700g/mL
Beilstein 17, V,3, 374
Refractive Index 1.4770 to 1.4800 (20°C, 589nm)
Synonym DL-α-Methylfuran-2-methanol
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 1 LIGHT SENSITIVE
Stabilizer 0.3 to 0.5% hydroquinone
Storage Information Product may darken on storage
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 4208-64-4
Assay Percent Range 99%
Molecular Formula C6H8O2
Chemical Name or Material DL-1-(2-Furyl)ethanol
Name Note 99%
MDL Number MFCD00077775
Safety and Handling

LIGHT SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.