missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Divinyltetramethyldisiloxane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC249212500

 View more versions of this product

Catalog No. AC249212500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Divinyltetramethyldisiloxane
24°C
Glass bottle
0.813
Colorless
250g
96% min. (GC)
[H2C=CHSi(CH3)2]2O
1,1'-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, 1,3-divinyltetramethyldisiloxane, bisvinyltetramethyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, unii-sj4d71zqf4
BITPLIXHRASDQB-UHFFFAOYSA-N
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane
75819
Liquid
0.8130g/mL
Authentic
1.4110 to 1.413
139°C
-99.7°C
2627-95-4
C8H18OSi2
MFCD00014933
Solubility in water: insoluble
C[Si](C)(C=C)O[Si](C)(C)C=C
186.4
186.4
97%

Chemical Identifiers

2627-95-4
186.4
BITPLIXHRASDQB-UHFFFAOYSA-N
75819
C[Si](C)(C=C)O[Si](C)(C)C=C
C8H18OSi2
MFCD00014933
1,1'-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, 1,3-divinyltetramethyldisiloxane, bisvinyltetramethyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, unii-sj4d71zqf4
ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-099-6

SDS Documents