Learn More
Dithizone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1794718
Description
Dithizone acts as an indicator for metals. It is also used as a chelating agent for heavy metals like mercury and lead. It is also used to evaluate the purification of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDithizone acts as an indicator for metals. It is also used as a chelating agent for heavy metals like mercury and lead. It is also used to evaluate the purification of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes.
Solubility
Soluble in ethanol, chloroform, dimethylsulfoxide, and pyridine. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Dithizone | |
∼168°C (decomposition) | |
C13H12N4S | |
50 g | |
14,3377 | |
Soluble in ethanol,chloroform,dimethylsulfoxide,and pyridine. Insoluble in water. | |
S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1 | |
256.33 | |
256.33 |
60-10-6 | |
Odorless | |
MFCD00003025 | |
748838 | |
dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone | |
UOFGSWVZMUXXIY-UHFFFAOYSA-N | |
1-anilino-3-phenyliminothiourea | |
657262 | |
98% |
Chemical Identifiers
60-10-6 | |
256.33 | |
UOFGSWVZMUXXIY-UHFFFAOYSA-N | |
657262 |
C13H12N4S | |
MFCD00003025 | |
dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone | |
S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-454-1
RTECSNumber : LQ9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only