Please login to your online account to display your discounted pricing

Alfa Aesar™ Dithiobiuret, 97%

$415.61 - $415.61

Chemical Identifiers

Chemical Name or Material Dithiobiuret
CAS 541-53-7
Molecular Formula C2H5N3S2
Formula Weight 135.21
MDL Number MFCD00037832
UN Number UN2811
Beilstein 1756621
Merck Index 14,3371
Synonym Dicarbonodithioimidic diamide; Thioimidodicarbonic diamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1617506

 
alfa aesar™
A1617506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1617514

 
alfa aesar™
A1617514
25g Each for $415.61
Description & Specifications

Specifications

Formula Weight 135.21
Chemical Name or Material Dithiobiuret
TSCA TSCA
Assay 97%
Molecular Formula C2H5N3S2
MDL Number MFCD00037832
CAS 541-53-7
UN Number UN2811
Beilstein 1756621
Merck Index 14,3371
Synonym Dicarbonodithioimidic diamide; Thioimidodicarbonic diamide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3 GHS P Statement
P280h-P270-P301+P310
Wear protective gloves/clothing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number EC1575000
Odor Odorless
Melting Point ≈183°C (dec.)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280h-P270-P301+P310
Wear protective gloves/clothing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H300
Fatal if swallowed.