Corning™ Dissolvable Microcarriers
Corning™ Dissolvable Microcarriers are designed to simplify cell harvest, separation and concentration processes and are ideal for bioprocess scale-up.
$472.82 - $472.82
Specifications
Product Type Dissolvable microcarrier
Culture Area 5000cm2/g
Particle Size (Metric) 200-300μm fully hydrated
Material Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions
View More Specs
Products 1
Catalog Number Mfr. No. Surface Treatment Quantity Price Quantity  
Catalog Number Mfr. No. Surface Treatment Quantity Price Quantity  
07-201-122 View Documents Corning™
4987
Corning™ Synthemax II 1g Case for $472.82
Description
Description
  • Offer cell separations that are faster, gentler, and more convenient than other standard commercially available microcarriers
  • Comprised of polygalacturonic acid (PGA) polymer chains cross linked via calcium ions pre-coated with either denatured collagen or Corning Synthemax™ II to enhance cell attachment for optimal yield and viability
  • Dissolvable within 10 to 20 minutes using EDTA and pectinase for easy separation of cells from the actual microcarriers
  • Narrow size distribution (1.02-1.03 g/cm3 density, 200-300μm fully hydrated) and large surface area (5000cm2/g) result in a more homogenous culture
  • Optical clarity for visualizing cell morphology
  • Presterilized to Sterility Assurance Level (SAL) of 10-6
  • Nonpyrogenic
  • Applications areas include cell therapy, vaccine production or large-volume adherent cell culture scale-up and cell banking
Specifications
Specifications
Dissolvable microcarrier
5000cm2/g
200-300μm fully hydrated
Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions