Honeywell™ Dissection Face Shield
North face shield replacement visors with a universal fit to help meet the most demanding applications.
Manufacturer:  Honeywell™ A815240
Catalog No. 29-634-259
Description
Description
High impact-rated clear polycarbonate is an easy-to-store die-cut visor, sized 8 x 15.5 x 0.04 in.
Specifications
Specifications
8 in.
Universal
15.5 in.
10/Pk.
Clear