Dissecting Needles, Probes, and Malls
Help students master dissecting techniques — choose the right needle and probe for the job.
Manufacturer:  Fisher Science Education 0136CH
 View more versions of this product
Catalog No. S17380
Description
Description
Our selection of dissecting needles and probes includes all high-quality stainless-steel instruments with precision points for precision work.
Teasing Needle with Metal Holder
  • 5cm long needle holder with an adjustable chuck for tightening or replacing the needles
  • Includes one straight needle
  • Replacement needles available separately (Bent — S17340B and Straight — S17340S, not pictured)
Flexible Probe with Eye
  • One end of this 152mm probe is olive-tapered
  • Other end is flat with an eye
Seeker Mall Probe
  • 152mm mall probe fashioned from chrome-plated steel with a hexagonal handle
  • One end is tapered to an angled point
  • Other end is blunt
Probe and Hook
  • Eight-sided handle on probe for easy turning and gripping while dissecting
  • Curved tenaculum end has a sharpened inside edge for cutting
  • Straight, tapered point is also sharp
Huber Probe
  • Double-ended probe and seeker
  • Has angular, tapered probe on one end
  • Other end has straight, tapered probe
Specifications
Specifications
Precision work, help students master dissecting techniques
6 in.
Stainless Steel
Hexagonal handle
15.24cm
Seeker Mall Probe