missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Disposable Polystyrene Weighing Dishes
Easy-grip contour sides for nonslip handling.
Manufacturer: Fisher Science Education WB316100
Description
Perfect for staining or destaining electrophoresis mini-gels.
- A pouring spout can be formed by squeezing
- Shape ensures its flexibility on a balance platform
- Smooth surface and rounded corners make powdered or granular samples easy to remove
Specifications
Disposable | |
Square | |
Perfect for staining or destaining electrophoresis mini-gels | |
3.3 in. | |
3.3 in. | |
1.9 cm |
Polystyrene | |
Weighing Dish | |
3.3 x 3.3 x 0.75 in. (8.3 x 8.3 x 1.9 cm) | |
8.3 cm | |
8.3 cm | |
0.75 in. |