missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Disposable PES Filter Units

Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.

Supplier:  Fisherbrand™ FB12566503

 View more versions of this product

Catalog No. FB12566503



Includes: Individually packaged sterile screw top style bottle cap

Description

Description

Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.

  • PES membrane is low protein binding to maintain protein balance
  • PES membrane has low extractables to maintain media purity
  • Upper chamber of filter units is clearly marked with pore size, membrane type, catalog number, lot number and expiration date 
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
  • 0.2μm units for sterile filtration
  • 0.45μm units for fluid clarification and particle removal

Certifications

Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility

Specifications

Specifications

Low binding
Single-use
PES
Sterile
Filter Unit
50 mm
Non-pyrogenic per USP Standard Lot-to-Lot
PES
0.45 μm
Fluid clarification and particle removal
250 mL
SDS
Documents

Documents

Promotions

Promotions