missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Disposable IR Cards, 5x10cm, NaCl window, 15mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Supplier: Thermo Scientific™ 43930KJ
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Size: 5 x 10cm
- NaCl window
- 15mm aperture
Specifications
Disposable IR Cards | |
5 Pk. |